1-(1-Methyl-1H-pyrazol-4-yl)ethanamine hydrochloride 95%

Lieferant: Apollo Scientific

OR920887-1G
APOSOR920887-1GEA 522 EUR
APOSOR920887-1G
1-(1-Methyl-1H-pyrazol-4-yl)ethanamine hydrochloride 95%
1-(1-Methyl-1H-pyrazol-4-yl)ethanamine hydrochloride
Formel: C₆H₁₁N₃·ClH
Molecular Weight: 161,633 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD22199336
CAS-Nummer: 1396762-20-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR