N-(2-Oxo-2-phenylethyl)formamide 95%

Lieferant: Apollo Scientific

OR920894-1G
APOSOR920894-1GEA 311 EUR
APOSOR920894-1G
N-(2-Oxo-2-phenylethyl)formamide 95%
N-(2-Oxo-2-phenylethyl)formamide
Formel: C₉H₉NO₂
Molecular Weight: 163.173 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21121748
CAS-Nummer: 73286-37-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR