4,4'-(1,2-Ethandiyl)dipyridin 95%

Lieferant: Apollo Scientific

OR920918-5G
APOSOR920918-5GEA 82.8 EUR
APOSOR920918-5G
4,4'-(1,2-Ethandiyl)dipyridin 95%
1,2-Di(4-pyridyl)ethane
Formel: C₁₂H₁₂N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00006451
CAS-Nummer: 4916-57-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR