2-Oxotetrahydrofuran-3-yl methacrylate 95%
Lieferant: Apollo Scientific
Synonyme:
GBLMA, 2-Methylacrylic acid 2-Oxotetrahydrofuran-3-yl ester
OR920934-1G
APOSOR920934-1GEA
609
EUR
APOSOR920934-1G
2-Oxotetrahydrofuran-3-yl methacrylate 95%
2-Oxotetrahydrofuran-3-yl methacrylate
Formel:
C₈H₁₀O₄ Molecular Weight: 170,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD19689400 CAS-Nummer: 195000-66-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...