Pyridin-4-yl(1H-pyrrol-2-yl)methanol 95%

Lieferant: Apollo Scientific

OR920984-1G
APOSOR920984-1GEA 538 EUR
APOSOR920984-1G
Pyridin-4-yl(1H-pyrrol-2-yl)methanol 95%
Pyridin-4-yl(1H-pyrrol-2-yl)methanol
Formel: C₁₀H₁₀N₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD21365097
CAS-Nummer: 1359828-89-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR