2-(3-Chloropyridin-4-yl)propan-2-ol 95%

Lieferant: Apollo Scientific

OR921005-1G
APOSOR921005-1GEA 683 EUR
APOSOR921005-1G
2-(3-Chloropyridin-4-yl)propan-2-ol 95%
2-(3-Chloropyridin-4-yl)propan-2-ol
Formel: C₈H₁₀ClNO
Molecular Weight: 171,624 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD19690066
CAS-Nummer: 77332-83-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR