S-Sodium ethanethiosulphonate 95%
Lieferant: Apollo Scientific
OR921031-100G
OR921031-25G
OR921031-5G
APOSOR921031-100GEA
429
EUR
APOSOR921031-100G
APOSOR921031-25G
APOSOR921031-5G
S-Sodium ethanethiosulphonate 95%
S-Sodium ethanethiosulphonate
Formel:
C₂H₆O₂S₂ Lagertemperatur: Raumtemperatur |
CAS-Nummer:
31999-88-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...