4-Nitrosoresorcinol-1-monomethyl ether 95%

Lieferant: Apollo Scientific

OR921045-1G OR921045-25G
APOSOR921045-1GEA 0 EUR
APOSOR921045-1G APOSOR921045-25G
4-Nitrosoresorcinol-1-monomethyl ether 95%
4-Nitrosoresorcinol-1-monomethyl ether
Formel: C₇H₇NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00059952
CAS-Nummer: 13895-38-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR