4-Nitrosoresorcinol-1-monomethyl ether 95%
Lieferant: Apollo Scientific
OR921045-1G
OR921045-25G
APOSOR921045-1GEA
0
EUR
APOSOR921045-1G
APOSOR921045-25G
4-Nitrosoresorcinol-1-monomethyl ether 95%
4-Nitrosoresorcinol-1-monomethyl ether
Formel:
C₇H₇NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00059952 CAS-Nummer: 13895-38-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...