2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol

Lieferant: Apollo Scientific

Synonyme: 2-((5-Bromopyridin-2-yl)diazenyl)-5-(diethylamino)phenol, 2-(4-Diethylamino-2-hydroxyphenylazo)-5-bromopyridine

OR921049-1G
APOSOR921049-1GEA 111 EUR
APOSOR921049-1G
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol
2-(5-Bromo-2-pyridylazo)-5-(diethylamino)phenol
Formel: C₁₅H₁₇BrN₄O
Molecular Weight: 349.23 g/mol
Schmelzpunkt: 157…158 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00006255
CAS-Nummer: 14337-53-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR