p-Xylylendiamin 99%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 1,4-Bis(aminomethyl)benzene, 1,4-Phenylenedimethanamine

OR921104-25G
APOSOR921104-25GEA 108 EUR
APOSOR921104-25G
p-Xylylendiamin 99%
p-Xylylendiamin
Formel: C₈H₁₂N₂
Molecular Weight: 136.20 g/mol
Siedepunkt: 230 °C/10mmHg
Schmelzpunkt: 59…63 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00009821
CAS-Nummer: 539-48-0
UN: 3259
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR