6-Chloro-[1,2,4]triazolo[1,5-a]pyridine 95%

Lieferant: Apollo Scientific

OR921114-1G
APOSOR921114-1GEA 383 EUR
APOSOR921114-1G
6-Chloro-[1,2,4]triazolo[1,5-a]pyridine 95%
6-Chloro-[1,2,4]triazolo[1,5-a]pyridine
Formel: C₆H₄ClN₃
Molecular Weight: 153,569 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD23708542
CAS-Nummer: 1427357-23-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR