1-Indanone-4-carboxylic acid 96%

Lieferant: Apollo Scientific

OR921127-1G OR921127-250MG
APOSOR921127-1GEA 632 EUR
APOSOR921127-1G APOSOR921127-250MG
1-Indanone-4-carboxylic acid 96%
1-Indanone-4-carboxylic acid
Formel: C₁₀H₈O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD03411485
CAS-Nummer: 56461-20-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR