4-Methoxycinnamonitrile (cis- and trans- mixture) 95%

Lieferant: Apollo Scientific
Achtung

OR921138-5G
APOSOR921138-5GEA 95.7 EUR
APOSOR921138-5G
4-Methoxycinnamonitrile (cis- and trans- mixture) 95%
4-Methoxycinnamonitrile (cis- and trans- mixture)
Formel: C₁₀H₉NO
Siedepunkt: 127…140 °C/1mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00001934
CAS-Nummer: 28446-68-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR