(3-Chlorphenyl)methylsulfon 95%

Lieferant: Apollo Scientific

Synonyme: 1-Chlor-3-(methylsulfonyl)benzol

OR921141-1G
APOSOR921141-1GEA 342 EUR
APOSOR921141-1G
(3-Chlorphenyl)methylsulfon 95%
(3-Chlorphenyl)methylsulfon
Formel: C₇H₇ClO₂S
Lagertemperatur: Raumtemperatur
CAS-Nummer: 21383-00-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR