4-Hydroxy-3-methoxypropiophenone 95%
Lieferant: Apollo Scientific
Synonyme:
1-(4-Hydroxy-3-methoxyphenyl)propan-1-one
OR921143-1G
APOSOR921143-1GEA
66.7
EUR
APOSOR921143-1G
4-Hydroxy-3-methoxypropiophenone 95%
4-Hydroxy-3-methoxypropiophenone
Formel:
C₁₀H₁₂O₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00482070 CAS-Nummer: 1835-14-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...