Phenaceturic acid 98%

Lieferant: Apollo Scientific

Synonyme: 2-(2-Phenylacetamido)acetic acid, N-(Phenylacetyl)glycine

OR921146-25G
APOSOR921146-25GEA 125 EUR
APOSOR921146-25G
Phenaceturic acid 98%
Phenaceturic acid
Formel: C₁₀H₁₁NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00021744
CAS-Nummer: 500-98-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR