[4-(Thien-3-yl)phenyl]methylamine 97%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 3-[4-(Aminomethyl)phenyl]thiophene, 4-(Thien-3-yl)benzylamine

OR9212-1G
APOSOR9212-1GEA 282 EUR
APOSOR9212-1G
[4-(Thien-3-yl)phenyl]methylamine 97%
[4-(Thien-3-yl)phenyl]methylamine
Formel: C₁₁H₁₁NS
Molecular Weight: 189,277 g/mol
Schmelzpunkt: 116…117 °C
Lagertemperatur: Kühlschrank
MDL: MFCD08271959
CAS-Nummer: 876317-18-9
UN: 3259
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR