sec-Butyl methanesulphonate 96%
Lieferant: Apollo Scientific
OR921213-1G
OR921213-5G
APOSOR921213-1GEA
270
EUR
APOSOR921213-1G
APOSOR921213-5G
sec-Butyl methanesulphonate 96%
sec-Butyl methanesulphonate
Formel:
C₅H₁₂O₃S Siedepunkt: 82 °C (2 mm Hg) Dichte: 1,1053 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD01715284 CAS-Nummer: 16156-54-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...