5,6,7,8-Tetrahydro-imidazo[1,2-a]pyridine-6-carboxylic acid methyl ester 95%

Lieferant: Apollo Scientific

OR921219-1G
APOSOR921219-1GEA 647 EUR
APOSOR921219-1G
5,6,7,8-Tetrahydro-imidazo[1,2-a]pyridine-6-carboxylic acid methyl ester 95%
5,6,7,8-Tetrahydro-imidazo[1,2-a]pyridine-6-carboxylic acid methyl ester
Formel: C₉H₁₂N₂O₂
Molecular Weight: 180,204 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11849321
CAS-Nummer: 139183-98-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR