(4-Bromothiazol-2-yl)methanol 96%
Lieferant: Apollo Scientific
OR921317-1G
APOSOR921317-1GEA
273
EUR
APOSOR921317-1G
(4-Bromothiazol-2-yl)methanol 96%
(4-Bromothiazol-2-yl)methanol
Formel:
C₄H₄BrNOS Molecular Weight: 194.05 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD09746337 CAS-Nummer: 204513-31-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...