Methyl (2r)-2-methylpyrrolidine-2-carboxylate hydrochloride 95%

Lieferant: Apollo Scientific

OR921329-1G OR921329-250MG
APOSOR921329-1GEA 301 EUR
APOSOR921329-1G APOSOR921329-250MG
Methyl (2r)-2-methylpyrrolidine-2-carboxylate hydrochloride 95%
Methyl (2r)-2-methylpyrrolidine-2-carboxylate hydrochloride
Formel: C₇H₁₃NO₂·ClH
Molecular Weight: 179.645 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD20685906
CAS-Nummer: 109837-32-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR