Methyl (2r)-2-methylpyrrolidine-2-carboxylate hydrochloride 95%
Lieferant: Apollo Scientific
OR921329-1G
OR921329-250MG
APOSOR921329-1GEA
301
EUR
APOSOR921329-1G
APOSOR921329-250MG
Methyl (2r)-2-methylpyrrolidine-2-carboxylate hydrochloride 95%
Methyl (2r)-2-methylpyrrolidine-2-carboxylate hydrochloride
Formel:
C₇H₁₃NO₂·ClH Molecular Weight: 179.645 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD20685906 CAS-Nummer: 109837-32-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...