2-(1-(Ethylsulphonyl)azetidin-3-ylidene)acetonitrile 97%
Lieferant: Apollo Scientific
OR921330-100G
APOSOR921330-100GEA
186
EUR
APOSOR921330-100G
2-(1-(Ethylsulphonyl)azetidin-3-ylidene)acetonitrile 97%
2-(1-(Ethylsulphonyl)azetidin-3-ylidene)acetonitrile
Formel:
C₇H₁₀N₂O₂S Molecular Weight: 186.232 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD22987586 CAS-Nummer: 1187595-85-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...