3-Hydroxycyclobutan-1-one

Lieferant: Apollo Scientific

OR921380-1G
APOSOR921380-1GEA 649 EUR
APOSOR921380-1G
3-Hydroxycyclobutan-1-one
3-Hydroxycyclobutan-1-one
Formel: C₄H₆O₂
Molecular Weight: 86.089 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21604157
CAS-Nummer: 15932-93-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR