Ethyl (2R)-2-methoxypropanoate 95%

Lieferant: Apollo Scientific

Synonyme: (R)-Ethyl 2-methoxypropanoate

OR921409-1G
APOSOR921409-1GEA 213 EUR
APOSOR921409-1G
Ethyl (2R)-2-methoxypropanoate 95%
Ethyl (2R)-2-methoxypropanoate
Formel: C₆H₁₂O₃
Molecular Weight: 132,158 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01632583
CAS-Nummer: 40105-20-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR