Benzoyl-DL-alanine 98%

Lieferant: Apollo Scientific

OR921412-100G
APOSOR921412-100GEA 104 EUR
APOSOR921412-100G
Benzoyl-DL-alanine 98%
Benzoyl-DL-alanine
Formel: C₁₀H₁₁NO₃
Molecular Weight: 193.20 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00020393
CAS-Nummer: 1205-02-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR