1-Brom-2-propanol 95%

Lieferant: Apollo Scientific

OR921430-25G
APOSOR921430-25GEA 227 EUR
APOSOR921430-25G
1-Brom-2-propanol 95%
1-Brom-2-propanol
Formel: BrH₂CCH(OH)CH₃
Molecular Weight: 138,99 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00004529
CAS-Nummer: 19686-73-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR