1-Butyl-1-phenylhydrazin 95%

Lieferant: Apollo Scientific

OR921493-1G OR921493-25G OR921493-5G
APOSOR921493-1GEA 161 EUR
APOSOR921493-1G APOSOR921493-25G APOSOR921493-5G
1-Butyl-1-phenylhydrazin 95%
1-Butyl-1-phenylhydrazine
Formel: C₁₀H₁₆N₂
Lagertemperatur: Raumtemperatur
MDL: MFCD03844777
CAS-Nummer: 61715-75-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR