Methyl[(1-methyl-1H-pyrazol-4-yl)methyl]amine dihydrochloride 95%

Lieferant: Apollo Scientific

OR921502-1G
APOSOR921502-1GEA 522 EUR
APOSOR921502-1G
Methyl[(1-methyl-1H-pyrazol-4-yl)methyl]amine dihydrochloride 95%
Methyl[(1-methyl-1H-pyrazol-4-yl)methyl]amine dihydrochloride
Formel: C₆H₁₁N₃·2ClH
Lagertemperatur: Raumtemperatur
MDL: MFCD25371024
CAS-Nummer: 1431965-92-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR