3-Amino-6-cyano-2-methylpyridine 95%

Lieferant: Apollo Scientific

Synonyme: 5-Amino-6-methylpicolinonitrile

OR921514-1G OR921514-250MG
APOSOR921514-1GEA 59.6 EUR
APOSOR921514-1G APOSOR921514-250MG
3-Amino-6-cyano-2-methylpyridine 95%
3-Amino-6-cyano-2-methylpyridine
Formel: C₇H₇N₃
Molecular Weight: 133.154 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD08062828
CAS-Nummer: 1079054-78-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR