L(-)-Menthol 95%
Lieferant: Apollo Scientific
Synonyme:
L(-)-Menthylalkoholol, (1R)-(-)-Menthol, Levomenthol, (1R,2S,5R)-(-)-Menthol, (1α,2β,4β)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexan, (1S,2R,4R)-(-)-2-Hydroxy-1-isopropyl-4-methylcyclohexan
OR921562-500G
APOSOR921562-500GEA
77
EUR
APOSOR921562-500G
L(-)-Menthol 95%
L(-)-Menthol
Formel:
C₁₀H₂₀O Molecular Weight: 156.27 g/mol Siedepunkt: 212 Flammpunkt: 96 Lagertemperatur: Raumtemperatur |
MDL:
MFCD00062979 CAS-Nummer: 2216-51-5 Merck Index: 13,05861 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...