Methyl 3-(4-cyanophenyl)propanoate 95%

Lieferant: Apollo Scientific

OR921564-1G
APOSOR921564-1GEA 364 EUR
APOSOR921564-1G
Methyl 3-(4-cyanophenyl)propanoate 95%
Methyl 3-(4-cyanophenyl)propanoate
Formel: C₁₁H₁₁NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD19440756
CAS-Nummer: 75567-85-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR