Methyl-6-hydroxy-2-naphthoat 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Hydroxy-2-naphthoic acid methyll ester, Methyl 6-hydroxy-2-naphthoate

OR921588-25G
APOSOR921588-25GEA 91.5 EUR
APOSOR921588-25G
Methyl-6-hydroxy-2-naphthoat 97%
Methyl-6-hydroxy-2-naphthoat
Formel: C₁₂H₁₀O₃
Molecular Weight: 202.2060 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00100526
CAS-Nummer: 17295-11-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR