3-(Ethylamino)phenol 95%

Lieferant: Apollo Scientific

OR921595-10G OR921595-1G OR921595-5G
APOSOR921595-10GEA 754 EUR
APOSOR921595-10G APOSOR921595-1G APOSOR921595-5G
3-(Ethylamino)phenol 95%
3-(Ethylamino)phenol
Formel: C₈H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD00059617
CAS-Nummer: 621-31-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR