Methyl 5-(methylamino)nicotinate 95%

Lieferant: Apollo Scientific

OR921634-1G OR921634-250MG
APOSOR921634-1GEA 632 EUR
APOSOR921634-1G APOSOR921634-250MG
Methyl 5-(methylamino)nicotinate 95%
Methyl 5-(methylamino)nicotinate
Formel: C₈H₁₀N₂O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD18256217
CAS-Nummer: 91702-86-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR