3-Chlorocyclobutane-1-carboxylic acid

Lieferant: Apollo Scientific

OR921690-1G OR921690-5G
APOSOR921690-1GEA 0 EUR
APOSOR921690-1G APOSOR921690-5G
3-Chlorocyclobutane-1-carboxylic acid
3-Chlorocyclobutane-1-carboxylic acid
Formel: C₅H₇ClO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD07366750
CAS-Nummer: 35207-71-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR