(R)-3-Methoxypiperidine hydrochloride 98%

Lieferant: Apollo Scientific

OR921704-1G
APOSOR921704-1GEA 130 EUR
APOSOR921704-1G
(R)-3-Methoxypiperidine hydrochloride 98%
(R)-3-Methoxypiperidine hydrochloride
Formel: C₆H₁₃NO·ClH
Molecular Weight: 151,634 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD17015899
CAS-Nummer: 688809-95-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR