7-Methoxy-6-methylquinoline 95%

Lieferant: Apollo Scientific

OR921709-1G
APOSOR921709-1GEA 0 EUR
APOSOR921709-1G
7-Methoxy-6-methylquinoline 95%
7-Methoxy-6-methylquinoline
Formel: C₁₁H₁₁NO
Lagertemperatur: Raumtemperatur
MDL: MFCD18452204
CAS-Nummer: 97581-31-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR