Diisopropylsulfat 95%
Lieferant: Apollo Scientific
OR921753-10G
OR921753-1G
OR921753-5G
APOSOR921753-10GEA
712
EUR
APOSOR921753-10G
APOSOR921753-1G
APOSOR921753-5G
Diisopropylsulfat 95%
Diisopropyl sulphate
Formel:
C₆H₁₄O₄S Lagertemperatur: Raumtemperatur |
MDL:
MFCD01682898 CAS-Nummer: 2973-10-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...