Diisopropylsulfat  95%

Lieferant: Apollo Scientific

OR921753-10G OR921753-1G OR921753-5G
APOSOR921753-10GEA 712 EUR
APOSOR921753-10G APOSOR921753-1G APOSOR921753-5G
Diisopropylsulfat  95%
Diisopropyl sulphate
Formel: C₆H₁₄O₄S
Lagertemperatur: Raumtemperatur
MDL: MFCD01682898
CAS-Nummer: 2973-10-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR