(S)-Cyclopropyl(phenyl)methanamine hydrochloride 99%
Lieferant: Apollo Scientific
|
Achtung
|
OR921766-250MG
APOSOR921766-250MGEA
84.1
EUR
APOSOR921766-250MG
(S)-Cyclopropyl(phenyl)methanamine hydrochloride 99%
(S)-Cyclopropyl(phenyl)methanamine hydrochloride
Formel:
C₁₀H₁₃N·ClH Molecular Weight: 183,678 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11101161 CAS-Nummer: 844470-80-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...