2-(Methylsulphonyl)thiophene 95%
Lieferant: Apollo Scientific
OR921801-5G
APOSOR921801-5GEA
154
EUR
APOSOR921801-5G
2-(Methylsulphonyl)thiophene 95%
2-(Methylsulphonyl)thiophene
Formel:
C₅H₆O₂S₂ Molecular Weight: 162.23 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052826 CAS-Nummer: 38695-60-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...