(4-Chloro-2-methylsulphanylpyrimidin-5-yl)methanol 95%

Lieferant: Apollo Scientific

Synonyme: (4-Chloro-2-(methylthio)pyrimidin-5-yl)methanol

OR921826-1G
APOSOR921826-1GEA 155 EUR
APOSOR921826-1G
(4-Chloro-2-methylsulphanylpyrimidin-5-yl)methanol 95%
(4-Chloro-2-methylsulphanylpyrimidin-5-yl)methanol
Formel: C₆H₇ClN₂OS
Molecular Weight: 190,65 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11858523
CAS-Nummer: 1044145-59-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR