Ethylsalicylat 98%
Lieferant: Apollo Scientific
Synonyme:
2-Hydroxybenzoesäureethylester, Ethyl-2-hydroxybenzoat, Salicylsäureethylester
OR921848-100G
APOSOR921848-100GEA
53.8
EUR
APOSOR921848-100G
Ethylsalicylat 98%
Ethylsalicylat
Formel:
C₉H₁₀O₃ Molecular Weight: 166,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00002215 CAS-Nummer: 118-61-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...