4-tert-Pentylcyclohexanon 95%

Lieferant: Apollo Scientific

OR921862-100G OR921862-25G
APOSOR921862-100GEA 143 EUR
APOSOR921862-100G APOSOR921862-25G
4-tert-Pentylcyclohexanon 95%
4-tert-Pentylcyclohexanon
Formel: C₁₁H₂₀O
Lagertemperatur: Raumtemperatur
MDL: MFCD00013798
CAS-Nummer: 16587-71-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR