Benzylacetat 95%

Lieferant: Apollo Scientific

OR921864-100G
APOSOR921864-100GEA 0 EUR
APOSOR921864-100G
Benzylacetat 95%
Benzylacetat
Formel: C₉H₁₀O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00008712
CAS-Nummer: 140-11-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR