(6-Chloropyridazin-3-yl)methanol

Lieferant: Apollo Scientific

OR921872-1G
APOSOR921872-1GEA 811 EUR
APOSOR921872-1G
(6-Chloropyridazin-3-yl)methanol
(6-Chloropyridazin-3-yl)methanol
Formel: C₅H₅ClN₂O
Molecular Weight: 144.559 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD11977451
CAS-Nummer: 1011487-94-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR