Mesitylglyoxylic acid 95%

Lieferant: Apollo Scientific

OR921873-5G
APOSOR921873-5GEA 137 EUR
APOSOR921873-5G
Mesitylglyoxylic acid 95%
Mesitylglyoxylic acid
Formel: C₁₁H₁₂O₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00151834
CAS-Nummer: 3112-46-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR