Ethyl(trimethylsilyl)acetat 95%
Lieferant: Apollo Scientific
Synonyme:
(Trimethylsilyl)essigsäureethylester
OR921875-100G
APOSOR921875-100GEA
88.6
EUR
APOSOR921875-100G
Ethyl(trimethylsilyl)acetat 95%
Ethyl(trimethylsilyl)acetat
Formel:
C₇H₁₆O₂Si Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009172 CAS-Nummer: 4071-88-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...